Assay Details
Target Gene Details
Entrez Gene ID: | 105377621 |
Gene Name: | uncharacterized LOC105377621 |
Gene Aliases: |
- |
Location: |
Chr.4:102418602-102502486 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105377621 | NR_136202.1 |
Target Gene Details
Entrez Gene ID: | 4790 |
Gene Name: | nuclear factor kappa B subunit 1 |
Gene Aliases: |
CVID12, EBP-1, KBF1, NF-kB1, NF-kappa-B, NF-kappaB, NFKB-p105, NFKB-p50, NFkappaB, p105, p50 |
Location: |
Chr.4:102501329-102617302 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFKB1 | NM_001165412.1 | NP_001158884.1 | ||
| NM_003998.3 | NP_003989.2 | |||
| BC051765.1 | AAH51765.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4442 | Chr.4:102498046 - 102529664 on Build GRCh38 | Insertion |
|
| esv27559 | Chr.4:102501056 - 102501786 on Build GRCh38 | Loss |
|
| nsv830020 | Chr.4:102466186 - 102660127 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map