Assay Details
Target Gene Details
Entrez Gene ID: | 29119 |
Gene Name: | catenin alpha 3 |
Gene Aliases: |
ARVD13, VR22 |
Location: |
Chr.10:65912518-67696217 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTNNA3 | NM_001127384.2 | NP_001120856.1 | ||
| NM_013266.3 | NP_037398.2 | |||
| XM_017016151.1 | XP_016871640.1 | |||
| XM_017016152.1 | XP_016871641.1 | |||
| XM_017016153.1 | XP_016871642.1 | |||
| XM_017016154.1 | XP_016871643.1 | |||
| XM_017016155.1 | XP_016871644.1 | |||
| XM_017016156.1 | XP_016871645.1 | |||
| AF091606.1 | AAF21801.1 | |||
| AK289756.1 | ||||
| AK308726.1 | ||||
| AK313882.1 | ||||
| BC065819.1 | AAH65819.1 |
Target Gene Details
Entrez Gene ID: | 347731 |
Gene Name: | leucine rich repeat transmembrane neuronal 3 |
Gene Aliases: |
- |
Location: |
Chr.10:66926034-67101551 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRTM3 | NM_178011.4 | 3 | 3550 | NP_821079.3 |
| NR_111909.1 | 3 | 1594 | ||
| BC111492.1 | 3 | 3502 | AAI11493.1 | |
| BX640611.1 | 3 | 3529 | CAE45717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520745 | Chr.10:66982077 - 67162484 on Build GRCh38 | Gain |
|
| dgv916n100 | Chr.10:66975498 - 67173658 on Build GRCh38 | Gain |
|
| dgv33e55 | Chr.10:66929809 - 67178536 on Build GRCh38 | Gain |
|
| nsv831905 | Chr.10:67096507 - 67239655 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map