Assay Details
Target Gene Details
Entrez Gene ID: | 56063 |
Gene Name: | transmembrane protein 234 |
Gene Aliases: |
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7 |
Location: |
Chr.1:32204807-32222371 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv300 | Chr.1:32206912 - 32241689 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map