Assay Details
Target Gene Details
Entrez Gene ID: | 617 |
Gene Name: | BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone |
Gene Aliases: |
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1 |
Location: |
Chr.2:218659656-218663443 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCS1L | NM_001079866.1 | NP_001073335.1 | ||
| NM_001257342.1 | NP_001244271.1 | |||
| NM_001257343.1 | NP_001244272.1 | |||
| NM_001257344.1 | NP_001244273.1 | |||
| NM_001318836.1 | NP_001305765.1 | |||
| NM_001320717.1 | NP_001307646.1 | |||
| NM_004328.4 | NP_004319.1 | |||
| XM_005246748.2 | 1 | 258 | XP_005246805.1 | |
| XM_005246749.3 | XP_005246806.1 | |||
| XM_006712678.1 | XP_006712741.1 | |||
| XM_017004631.1 | XP_016860120.1 | |||
| XM_017004632.1 | XP_016860121.1 | |||
| XM_017004633.1 | XP_016860122.1 | |||
| AF026849.1 | AAD08638.1 | |||
| AF038195.1 | AAB97365.1 | |||
| AK096210.1 | 1 | 312 | ||
| AK223518.1 | BAD97238.1 | |||
| AK289363.1 | ||||
| BC000416.2 | AAH00416.1 | |||
| BC007500.2 | AAH07500.1 | |||
| BP215082.1 | ||||
| BX571752.1 | CAE11877.1 | |||
| CB159298.1 | ||||
| DA540458.1 | ||||
| DA613959.1 | ||||
| EF672347.1 | ||||
| KP992846.1 |
More Information
Set Membership: |
|

Genomic Map