Assay Details
Target Gene Details
Entrez Gene ID: | 9289 |
Gene Name: | adhesion G protein-coupled receptor G1 |
Gene Aliases: |
BFPP, BPPR, GPR56, TM7LN4, TM7XN1 |
Location: |
Chr.16:57619535-57665039 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRG1 | NM_001145770.2 | 1 | 58 | NP_001139242.1 |
| NM_001290142.1 | NP_001277071.1 | |||
| NM_001290143.1 | NP_001277072.1 | |||
| NM_005682.6 | 1 | 58 | NP_005673.3 | |
| XM_005256240.4 | XP_005256297.1 | |||
| XM_005256242.4 | XP_005256299.1 | |||
| XM_005256249.3 | 1 | 53 | XP_005256306.1 | |
| XM_005256251.4 | XP_005256308.1 | |||
| XM_006721339.3 | XP_006721402.1 | |||
| XM_006721340.3 | XP_006721403.1 | |||
| XM_006721343.3 | XP_006721406.1 | |||
| XM_006721345.3 | XP_006721408.1 | |||
| XM_011523463.2 | XP_011521765.1 | |||
| XM_011523465.2 | XP_011521767.1 | |||
| AK075270.1 | ||||
| AK131550.1 | BAD18684.1 | |||
| AY358400.1 | AAQ88766.1 | |||
| CR936747.1 | ||||
| DA386131.1 | 1 | 58 | ||
| DB231209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833246 | Chr.16:57577216 - 57747868 on Build GRCh38 | Loss |
|
| nsv952037 | Chr.16:57619589 - 57621488 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map