Assay Details
Target Gene Details
Entrez Gene ID: | 57537 |
Gene Name: | sortilin related VPS10 domain containing receptor 2 |
Gene Aliases: |
- |
Location: |
Chr.4:7192607-7742837 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SORCS2 | NM_020777.2 | 1 | 402 | NP_065828.2 |
| XM_005247987.4 | 1 | 442 | XP_005248044.2 | |
| XM_011513514.2 | 1 | 442 | XP_011511816.1 | |
| XM_011513515.2 | 1 | 442 | XP_011511817.1 | |
| XM_011513516.2 | 1 | 442 | XP_011511818.1 | |
| XM_017008481.1 | 1 | 442 | XP_016863970.1 | |
| AF286190.1 | 1 | 301 | AAL04014.1 | |
| AI206155.1 | 1 | 304 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759221 | Chr.4:6986421 - 7268174 on Build GRCh38 | Gain |
|
| nsv829850 | Chr.4:7110782 - 7312289 on Build GRCh38 | Loss |
|
| dgv8942n54 | Chr.4:7151915 - 7195666 on Build GRCh38 | Loss |
|
| esv34033 | Chr.4:6971543 - 7388894 on Build GRCh38 | Loss |
|
| dgv2724n106 | Chr.4:7192373 - 7193073 on Build GRCh38 | Deletion |
|
| nsv593563 | Chr.4:7182257 - 7220635 on Build GRCh38 | Loss |
|
| nsv593561 | Chr.4:7153016 - 7218206 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map