Assay Details
Target Gene Details
Entrez Gene ID: | 100131390 |
Gene Name: | Sp9 transcription factor |
Gene Aliases: |
ZNF990 |
Location: |
Chr.2:174335093-174337540 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP9 | NM_001145250.1 | NP_001138722.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583666 | Chr.2:174324901 - 174346560 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map