Assay Details
Target Gene Details
Entrez Gene ID: | 81570 |
Gene Name: | ClpB homolog, mitochondrial AAA ATPase chaperonin |
Gene Aliases: |
ANKCLB, HSP78, MEGCANN, MGCA7, SKD3 |
Location: |
Chr.11:72292425-72434684 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLPB | NM_001258392.2 | 16 | 2641 | NP_001245321.1 |
| NM_001258393.2 | 15 | 2554 | NP_001245322.1 | |
| NM_001258394.2 | 18 | 2838 | NP_001245323.1 | |
| NM_030813.5 | 17 | 2731 | NP_110440.1 | |
| AK023214.1 | 16 | 2491 | BAB14467.1 | |
| BC006404.2 | 17 | 2571 | AAH06404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv385 | Chr.11:72270271 - 72306912 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75201671] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map