Assay Details
Target Gene Details
Entrez Gene ID: | 25983 |
Gene Name: | neuroguidin |
Gene Aliases: |
C14orf120, CANu1, LCP5, NGD, lpd-2 |
Location: |
Chr.14:23469689-23478193 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NGDN | NM_001042635.1 | NP_001036100.1 | ||
| NM_015514.1 | NP_056329.1 | |||
| AK022215.1 | ||||
| AK124318.1 | 3 | 3515 | ||
| AK291875.1 | ||||
| AL050003.1 | CAB43232.2 | |||
| BC030817.1 | AAH30817.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1040319 | Chr.14:23447084 - 23496308 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map