Assay Details
Target Gene Details
Entrez Gene ID: | 3931 |
Gene Name: | lecithin-cholesterol acyltransferase |
Gene Aliases: |
- |
Location: |
Chr.16:67939884-67944112 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCAT | NM_000229.1 | 2 | 232 | NP_000220.1 |
| BC014781.1 | 2 | 222 | AAH14781.1 | |
| BT009748.1 | 2 | 221 | AAP88750.1 | |
| M12625.1 | 2 | 488 | AAA59498.1 | |
| M26268.1 | 2 | 174 | AAA59499.1 | |
| X06537.1 | 2 | 324 | CAB56610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map