Assay Details
Target Gene Details
Entrez Gene ID: | 1355 |
Gene Name: | COX15, cytochrome c oxidase assembly homolog |
Gene Aliases: |
CEMCOX2 |
Location: |
Chr.10:99694793-99732667 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX15 | NM_001320974.1 | NP_001307903.1 | ||
| XM_006717634.3 | XP_006717697.1 | |||
| XM_017015715.1 | XP_016871204.1 |
Target Gene Details
Entrez Gene ID: | 57089 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 7 |
Gene Aliases: |
LALP1 |
Location: |
Chr.10:99659506-99711245 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENTPD7 | NM_020354.3 | 13 | 1856 | NP_065087.1 |
| AF269255.1 | 13 | 1821 | AAF90135.1 | |
| AK055540.1 | 13 | 1856 | ||
| AK309735.1 | 14 | 2039 | ||
| AK314542.1 | 13 | 1914 | ||
| BC119008.1 | 13 | 1822 | ||
| BC122857.1 | 13 | 1822 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7516 | Chr.10:99676637 - 99708437 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map