Assay Details
Target Gene Details
Entrez Gene ID: | 51585 |
Gene Name: | PCF11 cleavage and polyadenylation factor subunit |
Gene Aliases: |
- |
Location: |
Chr.11:83157095-83185794 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCF11 | NM_015885.3 | NP_056969.2 | ||
| XM_005274048.1 | 7 | 2406 | XP_005274105.1 | |
| XM_005274049.1 | 7 | 2406 | XP_005274106.1 | |
| XM_017017918.1 | XP_016873407.1 | |||
| AB020631.1 | ||||
| AF046935.1 | AAC03107.1 | |||
| AK025103.1 | ||||
| AK302606.1 | 7 | 2375 | ||
| BC065384.1 | ||||
| BC146778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33531 | Chr.11:82520885 - 83805659 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map