Assay Details
Target Gene Details
Entrez Gene ID: | 9867 |
Gene Name: | praja ring finger ubiquitin ligase 2 |
Gene Aliases: |
Neurodap1, RNF131 |
Location: |
Chr.5:109334709-109409974 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PJA2 | NM_014819.4 | 10 | 3772 | NP_055634.3 |
| XM_017010099.1 | 11 | 3848 | XP_016865588.1 | |
| AB007898.1 | 10 | 3669 | ||
| BC030826.1 | 10 | 3668 | AAH30826.1 | |
| CR749579.1 | 6 | 2129 | CAH18371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599338 | Chr.5:109193028 - 109340036 on Build GRCh38 | Gain |
|
| esv2763481 | Chr.5:109269689 - 109357995 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map