Assay Details
Target Gene Details
Entrez Gene ID: | 120071 |
Gene Name: | LARGE xylosyl- and glucuronyltransferase 2 |
Gene Aliases: |
GYLTL1B, PP5656 |
Location: |
Chr.11:45921621-45929096 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARGE2 | NM_001300721.1 | 13 | 1954 | NP_001287650.1 |
| NM_001300722.1 | 14 | 1881 | NP_001287651.1 | |
| NM_152312.4 | 13 | 1974 | NP_689525.3 | |
| XM_005252787.2 | 8 | 1234 | XP_005252844.1 | |
| XM_006718140.1 | 14 | 1861 | XP_006718203.1 | |
| XM_011519886.1 | 13 | 1935 | XP_011518188.1 | |
| XM_011519888.1 | 13 | 1805 | XP_011518190.1 | |
| XM_011519889.1 | 14 | 1861 | XP_011518191.1 | |
| XM_011519890.1 | 13 | 1805 | XP_011518192.1 | |
| XM_011519891.1 | XP_011518193.1 | |||
| XM_011519892.2 | 13 | 1935 | XP_011518194.1 | |
| XM_011519893.1 | XP_011518195.1 | |||
| XM_017017173.1 | 12 | 2028 | XP_016872662.1 | |
| AF258588.1 | 5 | 772 | AAG23791.1 | |
| AK055829.1 | 14 | 1881 | ||
| AK092526.1 | 9 | 1716 | BAC03909.1 | |
| AK096021.1 | 13 | 1950 | BAC04675.1 | |
| BC037291.2 | 13 | 1938 | AAH37291.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516442 | Chr.11:45878411 - 45961834 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map