Assay Details
Target Gene Details
Entrez Gene ID: | 729665 |
Gene Name: | coiled-coil domain containing 175 |
Gene Aliases: |
C14orf38, c14_5395 |
Location: |
Chr.14:59504212-59577276 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC175 | XM_011537120.2 | 20 | 3736 | XP_011535422.1 |
| XM_011537122.2 | 18 | 3513 | XP_011535424.1 | |
| XM_011537124.2 | 17 | 3360 | XP_011535426.1 |
Target Gene Details
Entrez Gene ID: | 51528 |
Gene Name: | JNK1/MAPK8-associated membrane protein |
Gene Aliases: |
C14orf100, CDA06, HSPC213, HSPC327, JAMP |
Location: |
Chr.14:59484443-59505407 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JKAMP | NM_001098625.2 | 7 | 1374 | NP_001092095.1 |
| NM_001284201.1 | 7 | 1915 | NP_001271130.1 | |
| NM_001284202.1 | 8 | 1501 | NP_001271131.1 | |
| NM_001284203.1 | 7 | 1424 | NP_001271132.1 | |
| NM_001284204.1 | 6 | 1315 | NP_001271133.1 | |
| NM_016475.4 | 7 | 1392 | NP_057559.2 | |
| XM_006720171.3 | 6 | 1291 | XP_006720234.1 | |
| XM_017021366.1 | 6 | 1224 | XP_016876855.1 | |
| AF151047.1 | 7 | 1314 | AAF36133.1 | |
| AF161445.1 | 7 | 1313 | AAF29005.1 | |
| AF212245.1 | 7 | 1314 | AAG41781.1 | |
| AF529304.1 | 5 | 1629 | AAP85636.1 | |
| AK021683.1 | 7 | 1273 | BAB13874.1 | |
| AK056145.1 | 7 | 1791 | ||
| AK128628.1 | 1 | 3198 | ||
| AL136696.1 | 7 | 1284 | CAB66631.1 | |
| BC005198.1 | 7 | 1300 | AAH05198.1 | |
| BC010359.1 | 7 | 1331 | AAH10359.1 | |
| BX161501.1 | 8 | 1384 | CAD61945.1 |
More Information
Set Membership: |
|

Genomic Map