Assay Details
Target Gene Details
Entrez Gene ID: | 137964 |
Gene Name: | glycerol-3-phosphate acyltransferase 4 |
Gene Aliases: |
1-AGPAT 6, AGPAT6, LPAAT-zeta, LPAATZ, TSARG7 |
Location: |
Chr.8:41578188-41625001 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPAT4 | NM_178819.3 | 2 | 722 | NP_848934.1 |
| XM_011544392.2 | 3 | 845 | XP_011542694.1 | |
| XM_011544393.1 | 2 | 734 | XP_011542695.1 | |
| XM_011544394.1 | 2 | 771 | XP_011542696.1 | |
| XM_017013042.1 | 3 | 872 | XP_016868531.1 | |
| XM_017013043.1 | 2 | 720 | XP_016868532.1 | |
| AF406612.1 | 2 | 710 | AAP21893.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831299 | Chr.8:41448617 - 41692486 on Build GRCh38 | Loss |
|
| esv3576565 | Chr.8:41064045 - 41790840 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map