Assay Details
Target Gene Details
Entrez Gene ID: | 4973 |
Gene Name: | oxidized low density lipoprotein receptor 1 |
Gene Aliases: |
CLEC8A, LOX1, LOXIN, SCARE1, SLOX1 |
Location: |
Chr.12:10158300-10172191 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OLR1 | NM_001172632.1 | NP_001166103.1 | ||
| NM_001172633.1 | NP_001166104.1 | |||
| NM_002543.3 | NP_002534.1 | |||
| AB010710.1 | BAA24580.1 | |||
| AB102861.1 | BAC81565.1 | |||
| AF035776.1 | AAC82329.1 | |||
| AK292124.1 | ||||
| AK295409.1 | ||||
| AK298040.1 | ||||
| AK308009.1 | ||||
| AK316353.1 | ||||
| AL551100.3 | ||||
| AX429248.1 | CAD36113.1 | |||
| BC022295.1 | AAH22295.1 | |||
| CB955704.1 | 1 | 118 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628528 | Chr.12:10157803 - 10176482 on Build GRCh38 | Loss |
|
| esv3628526 | Chr.12:10143326 - 10179529 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map