Assay Details
Target Gene Details
Entrez Gene ID: | 113091 |
Gene Name: | parathyroid hormone 2 |
Gene Aliases: |
TIP39 |
Location: |
Chr.19:49422414-49423441 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTH2 | NM_178449.3 | 2 | 366 | NP_848544.1 |
| AY037555.1 | 2 | 366 | AAK68689.1 | |
| AY048588.1 | AAL06597.1 | |||
| BC093995.1 | 2 | 305 | AAH93995.1 | |
| BC093997.1 | 2 | 309 | AAH93997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv52n68 | Chr.19:49286685 - 49456131 on Build GRCh38 | Loss |
|
| nsv953599 | Chr.19:49386444 - 49506743 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map