Assay Details
Target Gene Details
Entrez Gene ID: | 222537 |
Gene Name: | heparan sulfate-glucosamine 3-sulfotransferase 5 |
Gene Aliases: |
3-OST-5, 3OST5, HS3OST5, NBLA04021 |
Location: |
Chr.6:114055586-114342388 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HS3ST5 | NM_153612.3 | 2 | 391 | NP_705840.2 |
| XM_006715379.2 | 5 | 879 | XP_006715442.1 | |
| XM_011535588.2 | 3 | 1595 | XP_011533890.1 | |
| XM_017010470.1 | 5 | 1453 | XP_016865959.1 | |
| XM_017010471.1 | 5 | 2412 | XP_016865960.1 | |
| XM_017010472.1 | 4 | 2049 | XP_016865961.1 | |
| XM_017010473.1 | 5 | 1449 | XP_016865962.1 | |
| XM_017010474.1 | 5 | 887 | XP_016865963.1 | |
| AF503292.1 | 2 | 359 | AAN37737.1 | |
| AK126930.1 | 2 | 1051 | ||
| AK289907.1 | 5 | 878 | ||
| BC093911.1 | 2 | 392 | AAH93911.1 | |
| BC093913.1 | 2 | 392 | AAH93913.1 |
Target Gene Details
Entrez Gene ID: | 101927768 |
Gene Name: | uncharacterized LOC101927768 |
Gene Aliases: |
- |
Location: |
Chr.6:113969701-114340729 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927768 | NR_125845.1 |
More Information
Set Membership: |
|

Genomic Map