Assay Details
Target Gene Details
Entrez Gene ID: | 10365 |
Gene Name: | Kruppel like factor 2 |
Gene Aliases: |
LKLF |
Location: |
Chr.19:16324826-16328662 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF2 | NM_016270.3 | 2 | 959 | NP_057354.1 |
| AF134053.1 | 2 | 861 | AAD55891.1 | |
| AF205849.1 | 2 | 945 | AAF13295.1 | |
| AK225820.1 | 2 | 959 | ||
| BC071983.1 | 2 | 948 | AAH71983.1 | |
| BQ581981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833767 | Chr.19:16121391 - 16347760 on Build GRCh38 | Loss |
|
| nsv1118817 | Chr.19:16324689 - 16326389 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map