Assay Details
Target Gene Details
Entrez Gene ID: | 56255 |
Gene Name: | thioredoxin related transmembrane protein 4 |
Gene Aliases: |
DJ971N18.2, PDIA14, TXNDC13 |
Location: |
Chr.20:7977353-8019829 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMX4 | NM_021156.3 | 8 | 1208 | NP_066979.2 |
| AB032988.1 | 1 | 291 | BAA86476.1 | |
| AK074838.1 | 8 | 1140 | BAC11237.1 | |
| AK075404.1 | 8 | 1187 | BAC11599.1 | |
| AK226005.1 | 7 | 742 | ||
| AL118494.1 | 7 | 733 | CAB56344.1 | |
| AY359004.1 | 8 | 1125 | AAQ89363.1 | |
| BC033787.1 | 8 | 1019 | AAH33787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059059 | Chr.20:7793987 - 8053708 on Build GRCh38 | Gain |
|
| nsv585384 | Chr.20:7571617 - 8336371 on Build GRCh38 | Gain |
|
| esv2422203 | Chr.20:7968779 - 8463601 on Build GRCh38 | Duplication |
|
| esv3893279 | Chr.20:7582024 - 7999743 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map