Assay Details
Target Gene Details
Entrez Gene ID: | 91544 |
Gene Name: | UBX domain protein 11 |
Gene Aliases: |
COA-1, PP2243, SOC, SOCI, UBXD5 |
Location: |
Chr.1:26282282-26318265 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBXN11 | NM_001077262.1 | NP_001070730.1 | ||
| NM_145345.2 | NP_663320.2 | |||
| NM_183008.2 | NP_892120.2 | |||
| AF370425.1 | AAQ15261.1 | |||
| AF520989.1 | AAM76060.1 | |||
| AF521017.1 | AAM74201.1 | |||
| AF521018.1 | AAM74202.1 | |||
| AK074358.1 | ||||
| AK090792.1 | ||||
| AK093258.1 | BAC04112.1 | |||
| AK094198.1 | BAC04307.1 | |||
| AK097368.1 | BAC05024.1 | |||
| AY007163.1 | ||||
| BM987787.1 | ||||
| BX647774.1 | ||||
| BX648412.1 | CAH56155.1 | |||
| CR749413.1 | CAH18255.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745674 | Chr.1:26282147 - 26282585 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map