Assay Details
Target Gene Details
Entrez Gene ID: | 6506 |
Gene Name: | solute carrier family 1 member 2 |
Gene Aliases: |
EAAT2, GLT-1, HBGT |
Location: |
Chr.11:35251205-35420063 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC1A2 | NM_001195728.2 | 12 | 9505 | NP_001182657.1 |
| NM_001252652.1 | 12 | 9570 | NP_001239581.1 | |
| NM_004171.3 | 11 | 9867 | NP_004162.2 | |
| XM_005253067.1 | 11 | 9344 | XP_005253124.1 | |
| XM_011520285.2 | 11 | 9381 | XP_011518587.1 | |
| XM_017018136.1 | 11 | 10164 | XP_016873625.1 | |
| XM_017018137.1 | 11 | 9384 | XP_016873626.1 | |
| XM_017018138.1 | 11 | 9368 | XP_016873627.1 | |
| XM_017018139.1 | 10 | 9409 | XP_016873628.1 | |
| AK057674.1 | 1 | 431 | ||
| AY066021.1 | 11 | 9558 | AAL57716.1 | |
| BX538155.1 | 1 | 5377 | ||
| BX648750.1 | 1 | 5229 | ||
| CR626999.1 | 1 | 4013 | ||
| CR627009.1 | 1 | 3860 | ||
| CR627022.1 | 1 | 5879 | ||
| CR749842.1 | 1 | 5188 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042014 | Chr.11:34919190 - 37195907 on Build GRCh38 | Loss |
|
| nsv7733 | Chr.11:35243420 - 35276063 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map