Assay Details
Target Gene Details
Entrez Gene ID: | 1004 |
Gene Name: | cadherin 6 |
Gene Aliases: |
CAD6, KCAD |
Location: |
Chr.5:31193655-31329146 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH6 | NM_004932.3 | NP_004923.1 | ||
| XM_011513921.2 | XP_011512223.1 | |||
| XM_017008910.1 | XP_016864399.1 | |||
| XM_017008911.1 | XP_016864400.1 | |||
| AK291290.1 | ||||
| AU130185.1 | ||||
| BC000019.2 | AAH00019.1 | |||
| D31784.1 | BAA06562.1 | |||
| DC353791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5619n100 | Chr.5:31199804 - 31471735 on Build GRCh38 | Gain |
|
| esv3604572 | Chr.5:31134943 - 31452028 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map