Assay Details
Target Gene Details
Entrez Gene ID: | 9775 |
Gene Name: | eukaryotic translation initiation factor 4A3 |
Gene Aliases: |
DDX48, MUK34, NMP265, NUK34, RCPS, eIF4AIII |
Location: |
Chr.17:80135214-80147183 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4A3 | NM_014740.3 | NP_055555.1 | ||
| XM_011525522.1 | XP_011523824.1 | |||
| AK290608.1 | ||||
| BC003662.2 | AAH03662.1 | |||
| BC004386.1 | AAH04386.1 | |||
| BC011151.1 | AAH11151.1 | |||
| CA428254.1 | ||||
| CR456750.1 | CAG33031.1 | |||
| CR749455.1 | 2 | 2833 | ||
| D21853.1 | ||||
| X79538.1 | CAA56074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066606 | Chr.17:79955666 - 80224006 on Build GRCh38 | Gain |
|
| nsv469591 | Chr.17:80085322 - 80293486 on Build GRCh38 | Loss |
|
| nsv833561 | Chr.17:79977946 - 80167051 on Build GRCh38 | Loss |
|
| nsv833562 | Chr.17:80114523 - 80301339 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map