Assay Details
Target Gene Details
Entrez Gene ID: | 23125 |
Gene Name: | calmodulin binding transcription activator 2 |
Gene Aliases: |
- |
Location: |
Chr.17:4967992-4988297 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMTA2 | XM_006721478.3 | 1 | 163 | XP_006721541.1 |
| XM_006721481.3 | 1 | 163 | XP_006721544.1 |
Target Gene Details
Entrez Gene ID: | 388324 |
Gene Name: | inhibitor of CDK, cyclin A1 interacting protein 1 |
Gene Aliases: |
HSD45 |
Location: |
Chr.17:4988130-4997933 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INCA1 | NM_001167985.1 | NP_001161457.1 | ||
| NM_001167986.1 | NP_001161458.1 | |||
| NM_001167987.1 | NP_001161459.1 | |||
| NM_213726.2 | NP_998891.2 | |||
| XM_005256627.3 | XP_005256684.1 | |||
| XM_005256628.4 | XP_005256685.1 | |||
| XM_005256629.1 | XP_005256686.1 | |||
| XM_006721517.1 | XP_006721580.1 | |||
| XM_006721518.2 | XP_006721581.1 | |||
| XM_006721519.3 | XP_006721582.1 | |||
| XM_011523832.2 | XP_011522134.1 | |||
| AY601906.1 | AAT09152.1 | |||
| AY601907.1 | AAT09153.1 | |||
| AY604176.1 | AAT36739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833350 | Chr.17:4884713 - 5060776 on Build GRCh38 | Loss |
|
| nsv833349 | Chr.17:4853631 - 4991300 on Build GRCh38 | Loss |
|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| dgv5431n54 | Chr.17:4975665 - 5051911 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map