Assay Details
Target Gene Details
Entrez Gene ID: | 54880 |
Gene Name: | BCL6 corepressor |
Gene Aliases: |
ANOP2, MAA2, MCOPS2 |
Location: |
Chr.X:40051246-40177390 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
More Information
Set Membership: |
|

Genomic Map