Assay Details
Target Gene Details
Entrez Gene ID: | 259232 |
Gene Name: | sodium leak channel, non-selective |
Gene Aliases: |
CLIFAHDD, CanIon, IHPRF, IHPRF1, INNFD, VGCNL1, bA430M15.1 |
Location: |
Chr.13:101053774-101417206 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 46 - Exon 46 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NALCN | NM_052867.2 | NP_443099.1 | ||
| XM_011521067.2 | XP_011519369.1 | |||
| XM_011521068.2 | XP_011519370.1 | |||
| XM_011521069.2 | XP_011519371.1 | |||
| XM_017020534.1 | XP_016876023.1 | |||
| XM_017020535.1 | XP_016876024.1 | |||
| XM_017020536.1 | XP_016876025.1 | |||
| XM_017020537.1 | XP_016876026.1 | |||
| AK002089.1 | ||||
| AK055506.1 | ||||
| AK126621.1 | ||||
| AK126718.1 | ||||
| AK172752.1 | BAD18738.1 | |||
| AY141972.1 | AAN10255.1 |
Target Gene Details
Entrez Gene ID: | 100885778 |
Gene Name: | NALCN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:100708325-101059286 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NALCN-AS1 | NR_047687.1 | 8 | 1520 | |
| AK025806.1 | 8 | 1520 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1149 | Chr.13:101042454 - 101087249 on Build GRCh38 | Deletion |
|
| nsv832699 | Chr.13:100951779 - 101142880 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map