Assay Details
Target Gene Details
Entrez Gene ID: | 2067 |
Gene Name: | ERCC excision repair 1, endonuclease non-catalytic subunit |
Gene Aliases: |
COFS4, RAD10, UV20 |
Location: |
Chr.19:45407333-45478866 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833845 | Chr.19:45308814 - 45457371 on Build GRCh38 | Loss |
|
| nsv833846 | Chr.19:45381005 - 45469089 on Build GRCh38 | Loss |
|
| nsv428365 | Chr.19:45358027 - 45469089 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map