Assay Details
Target Gene Details
Entrez Gene ID: | 29964 |
Gene Name: | prickle planar cell polarity protein 4 |
Gene Aliases: |
C6orf49, OBTP, OEBT, TOMM6 |
Location: |
Chr.6:41780762-41787372 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRICKLE4 | NM_013397.5 | NP_037529.3 | ||
| BC110459.1 | AAI10460.1 | |||
| DA917216.1 |
More Information
Set Membership: |
|

Genomic Map