Assay Details
Target Gene Details
Entrez Gene ID: | 10645 |
Gene Name: | calcium/calmodulin dependent protein kinase kinase 2 |
Gene Aliases: |
CAMKK, CAMKKB |
Location: |
Chr.12:121237692-121298308 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560455 | Chr.12:121087788 - 121363295 on Build GRCh38 | Gain |
|
| nsv1054878 | Chr.12:121082077 - 121328424 on Build GRCh38 | Gain |
|
| nsv560454 | Chr.12:121019270 - 121307939 on Build GRCh38 | Loss |
|
| nsv952494 | Chr.12:121244498 - 121248497 on Build GRCh38 | Deletion |
|
| nsv560456 | Chr.12:121230451 - 121249371 on Build GRCh38 | Loss |
|
| nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map