Assay Details
Target Gene Details
Entrez Gene ID: | 50848 |
Gene Name: | F11 receptor |
Gene Aliases: |
CD321, JAM, JAM1, JAMA, JCAM, KAT, PAM-1 |
Location: |
Chr.1:160995211-161021343 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F11R | NM_016946.4 | NP_058642.1 | ||
| AF154005.1 | AAD43794.1 | |||
| AF172398.2 | AAD48877.1 | |||
| AF207907.1 | AAF22829.1 | |||
| AK026665.1 | ||||
| AK075152.1 | BAC11436.1 | |||
| AK299487.1 | ||||
| AK304412.1 | ||||
| AL136649.1 | CAB66584.1 | |||
| AY358896.1 | AAQ89255.1 | |||
| BC001533.2 | AAH01533.1 | |||
| BT020103.1 | AAV38906.1 | |||
| CR533512.1 | CAG38543.1 | |||
| DA907460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548058 | Chr.1:160955665 - 161019824 on Build GRCh38 | Loss |
|
| esv3578229 | Chr.1:161000223 - 161017227 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map