Assay Details
Target Gene Details
Entrez Gene ID: | 64333 |
Gene Name: | Rho GTPase activating protein 9 |
Gene Aliases: |
10C, RGL1 |
Location: |
Chr.12:57472255-57488814 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP9 | NM_001319850.1 | 1 | 153 | NP_001306779.1 |
| AK131415.1 | 1 | 67 | BAD18562.1 | |
| BC006107.1 | 1 | 153 | AAH06107.1 |
Target Gene Details
Entrez Gene ID: | 4141 |
Gene Name: | methionyl-tRNA synthetase |
Gene Aliases: |
CMT2U, ILFS2, ILLD, METRS, MRS, MTRNS, SPG70 |
Location: |
Chr.12:57487953-57516655 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARS | NM_004990.3 | NP_004981.2 | ||
| XM_006719398.3 | XP_006719461.1 | |||
| AK092986.1 | BAC04011.1 | |||
| AK122956.1 | ||||
| AK222767.1 | BAD96487.1 | |||
| AK290857.1 | ||||
| AK293945.1 | ||||
| AK303349.1 | ||||
| BC002384.2 | AAH02384.1 | |||
| BC006328.2 | AAH06328.1 | |||
| BC011639.2 | AAH11639.1 | |||
| BC011849.2 | AAH11849.1 | |||
| BC015011.1 | AAH15011.1 | |||
| BT007338.1 | AAP36002.1 | |||
| D84224.1 | BAA95668.1 | |||
| DA489707.1 | ||||
| X94754.1 | CAA64381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759901 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain |
|
| nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss |
|
| dgv2651n54 | Chr.12:57450266 - 57552694 on Build GRCh38 | Loss |
|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map