Assay Details
Target Gene Details
Entrez Gene ID: | 89953 |
Gene Name: | kinesin light chain 4 |
Gene Aliases: |
KNSL8, bA387M24.3 |
Location: |
Chr.6:43059594-43075099 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLC4 | NM_001289034.1 | NP_001275963.1 | ||
| NM_001289035.1 | NP_001275964.1 | |||
| NM_138343.3 | NP_612352.1 | |||
| NM_201521.2 | NP_958929.1 | |||
| NM_201522.2 | NP_958930.1 | |||
| NM_201523.2 | NP_958931.1 | |||
| XM_011514989.1 | XP_011513291.1 | |||
| XM_017011457.1 | XP_016866946.1 | |||
| AK055293.1 | ||||
| AK056393.1 | ||||
| AK094102.1 | ||||
| AK127018.1 | BAC86788.1 | |||
| AK127894.1 | BAC87179.1 | |||
| AK297433.1 | ||||
| BC012357.1 | AAH12357.1 | |||
| BC080637.1 | AAH80637.1 | |||
| BC103727.1 | AAI03728.1 | |||
| BQ688778.1 | ||||
| BX640717.1 | CAE45836.1 | |||
| DA782235.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462927 | Chr.6:42948182 - 43110718 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
| nsv462930 | Chr.6:43002769 - 43110718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map