Assay Details
Target Gene Details
Entrez Gene ID: | 10899 |
Gene Name: | jumping translocation breakpoint |
Gene Aliases: |
HJTB, HSPC222, PAR, hJT |
Location: |
Chr.1:153974269-153977975 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JTB | NM_006694.3 | NP_006685.1 | ||
| AB016488.1 | BAA33731.1 | |||
| AF115850.2 | AAD09822.2 | |||
| AF131797.1 | AAD20045.1 | |||
| AF151056.1 | 2 | 643 | AAF36142.1 | |
| AK312106.1 | ||||
| BC000499.1 | AAH00499.1 | |||
| BC000996.2 | AAH00996.1 | |||
| BC001363.1 | AAH01363.1 | |||
| BC001667.1 | AAH01667.1 | |||
| BC004239.1 | 3 | 596 | AAH04239.1 | |
| BC019277.1 | AAH19277.1 | |||
| BT007285.1 | AAP35949.1 | |||
| CR456985.1 | CAG33266.1 |
More Information
Set Membership: |
|

Genomic Map