Assay Details
Target Gene Details
Entrez Gene ID: | 30011 |
Gene Name: | SH3 domain containing kinase binding protein 1 |
Gene Aliases: |
CD2BP3, CIN85, GIG10, HSB-1, HSB1, MIG18 |
Location: |
Chr.X:19533965-19887626 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3KBP1 | NM_001024666.2 | 17 | 2481 | NP_001019837.1 |
| NM_001184960.1 | 13 | 1891 | NP_001171889.1 | |
| NM_031892.2 | 18 | 2793 | NP_114098.1 | |
| XM_005274494.2 | 19 | 2734 | XP_005274551.1 | |
| XM_011545498.2 | 20 | 2791 | XP_011543800.1 | |
| XM_011545499.2 | 19 | 2599 | XP_011543801.1 | |
| XM_011545500.2 | 19 | 2662 | XP_011543802.1 | |
| XM_011545502.2 | 13 | 1897 | XP_011543804.1 | |
| XM_017029459.1 | 19 | 2531 | XP_016884948.1 | |
| XM_017029460.1 | 19 | 2574 | XP_016884949.1 | |
| XM_017029461.1 | 18 | 2517 | XP_016884950.1 | |
| XM_017029462.1 | 18 | 2605 | XP_016884951.1 | |
| XM_017029463.1 | 17 | 2501 | XP_016884952.1 | |
| XM_017029464.1 | 19 | 2737 | XP_016884953.1 | |
| XM_017029465.1 | 18 | 2760 | XP_016884954.1 | |
| XM_017029466.1 | 17 | 2530 | XP_016884955.1 | |
| XM_017029467.1 | 18 | 2679 | XP_016884956.1 | |
| XM_017029468.1 | 17 | 2631 | XP_016884957.1 | |
| XM_017029469.1 | 13 | 1847 | XP_016884958.1 | |
| XM_017029470.1 | 12 | 1870 | XP_016884959.1 | |
| AF230904.1 | 18 | 2768 | AAF37854.1 | |
| AF542051.1 | 17 | 2444 | AAN77231.1 | |
| AY423734.1 | 19 | 2691 | AAS00497.1 | |
| BC015806.1 | 18 | 2695 | AAH15806.1 |
More Information
Set Membership: |
|

Genomic Map