Assay Details
Target Gene Details
Entrez Gene ID: | 351 |
Gene Name: | amyloid beta precursor protein |
Gene Aliases: |
AAA, ABETA, ABPP, AD1, APPI, CTFgamma, CVAP, PN-II, PN2 |
Location: |
Chr.21:25880550-26171128 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APP | NM_000484.3 | 18 | 3300 | NP_000475.1 |
| NM_001136016.3 | 17 | 3224 | NP_001129488.1 | |
| NM_001136129.2 | 15 | 2907 | NP_001129601.1 | |
| NM_001136130.2 | 17 | 3132 | NP_001129602.1 | |
| NM_001136131.2 | 16 | 2962 | NP_001129603.1 | |
| NM_001204301.1 | 17 | 3246 | NP_001191230.1 | |
| NM_001204302.1 | 16 | 3189 | NP_001191231.1 | |
| NM_001204303.1 | 15 | 3021 | NP_001191232.1 | |
| NM_201413.2 | 17 | 3243 | NP_958816.1 | |
| NM_201414.2 | 16 | 3075 | NP_958817.1 | |
| A02759.1 | 16 | 3021 | CAA00268.1 | |
| AF282245.1 | 15 | 2834 | AAQ14327.1 | |
| BC065529.1 | 17 | 3174 | AAH65529.1 | |
| BE907745.1 | 1 | 78 | ||
| BM876312.1 | 1 | 349 | ||
| CB854706.1 | 1 | 348 | ||
| M15533.1 | AAA35540.1 | |||
| M18734.1 | AAA51726.1 | |||
| S41243.1 | ||||
| X06989.1 | CAA30050.1 | |||
| Y00264.1 | 16 | 3022 | CAA68374.1 |
More Information
Set Membership: |
|

Genomic Map