Assay Details
Target Gene Details
Entrez Gene ID: | 3761 |
Gene Name: | potassium voltage-gated channel subfamily J member 4 |
Gene Aliases: |
HIR, HIRK2, HRK1, IRK-3, IRK3, Kir2.3 |
Location: |
Chr.22:38426327-38455199 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNJ4 | NM_004981.1 | 2 | 662 | NP_004972.1 |
| NM_152868.2 | 2 | 824 | NP_690607.1 | |
| BC032562.1 | 2 | 834 | AAH32562.1 | |
| BC113506.1 | 1 | 616 | ||
| BC113508.1 | 1 | 616 | ||
| CR456507.1 | 2 | 632 | CAG30393.1 | |
| S72503.1 | 2 | 663 | AAC60632.1 | |
| U07364.1 | 2 | 662 | AAA19962.1 | |
| U24056.1 | 2 | 799 | AAA66076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834195 | Chr.22:38300366 - 38490431 on Build GRCh38 | Loss |
|
| nsv1060077 | Chr.22:38419538 - 38460822 on Build GRCh38 | Gain |
|
| nsv817994 | Chr.22:38413480 - 38469858 on Build GRCh38 | Gain |
|
| nsv1058338 | Chr.22:38423344 - 38450414 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map