Assay Details
Target Gene Details
Entrez Gene ID: | 10134 |
Gene Name: | B-cell receptor-associated protein 31 |
Gene Aliases: |
6C6-AG, BAP31, CDM, DDCH, DXS1357E |
Location: |
Chr.X:153700492-153724746 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCAP31 | NM_001139441.1 | NP_001132913.1 | ||
| NM_001139457.2 | NP_001132929.1 | |||
| NM_001256447.1 | NP_001243376.1 | |||
| NM_005745.7 | NP_005736.3 | |||
| AK057613.1 | ||||
| AK125631.1 | ||||
| AK222494.1 | BAD96214.1 | |||
| AK223059.1 | BAD96779.1 | |||
| AU132745.1 | ||||
| BC014323.1 | AAH14323.1 | |||
| BC065292.1 | AAH65292.1 | |||
| BE729820.1 | ||||
| BG779815.1 | ||||
| BX474419.1 | ||||
| CX164660.1 | ||||
| DA134013.1 | ||||
| X81109.1 | CAA57015.1 | |||
| Z31696.1 | CAA83501.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| esv21656 | Chr.X:153687433 - 153750282 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map