Assay Details
Target Gene Details
Entrez Gene ID: | 84866 |
Gene Name: | transmembrane protein 25 |
Gene Aliases: |
- |
Location: |
Chr.11:118531088-118546670 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044281 | Chr.11:118503814 - 118554704 on Build GRCh38 | Gain |
|
| esv3627872 | Chr.11:118502100 - 118536457 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map