Assay Details
Target Gene Details
Entrez Gene ID: | 3594 |
Gene Name: | interleukin 12 receptor subunit beta 1 |
Gene Aliases: |
CD212, IL-12R-BETA1, IL12RB, IMD30 |
Location: |
Chr.19:18058994-18099027 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL12RB1 | NM_001290023.1 | 9 | 1048 | NP_001276952.1 |
| NM_001290024.1 | 10 | 1039 | NP_001276953.1 | |
| NM_005535.2 | 9 | 1048 | NP_005526.1 | |
| NM_153701.2 | 9 | 1048 | NP_714912.1 | |
| XM_006722741.3 | 10 | 1241 | XP_006722804.2 | |
| XM_011527966.2 | 10 | 1283 | XP_011526268.1 | |
| XM_011527967.2 | 10 | 1269 | XP_011526269.1 | |
| XM_011527968.2 | 10 | 1256 | XP_011526270.1 | |
| XM_011527969.2 | 10 | 1242 | XP_011526271.1 | |
| XM_011527970.2 | 10 | 1282 | XP_011526272.1 | |
| XM_011527971.2 | 10 | 1280 | XP_011526273.1 | |
| XM_011527972.2 | 10 | 1280 | XP_011526274.1 | |
| XM_011527973.2 | 9 | 1074 | XP_011526275.1 | |
| XM_011527974.2 | 9 | 1062 | XP_011526276.1 | |
| XM_011527975.2 | 10 | 1239 | XP_011526277.1 | |
| XM_011527976.2 | 10 | 1280 | XP_011526278.1 | |
| XM_011527977.2 | 10 | 1232 | XP_011526279.1 | |
| XM_017026762.1 | 5 | 420 | XP_016882251.1 | |
| AK225919.1 | 9 | 979 | ||
| AK290423.1 | 9 | 976 | ||
| BC029121.1 | 9 | 937 | AAH29121.1 | |
| BC137404.1 | 9 | 893 | ||
| BC137406.1 | 9 | 893 | ||
| BX647221.1 | 10 | 1039 | ||
| U03187.1 | 9 | 932 | AAA21340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470130 | Chr.19:17936474 - 18123777 on Build GRCh38 | Loss |
|
| nsv833773 | Chr.19:18042161 - 18192178 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map