Assay Details
Target Gene Details
Entrez Gene ID: | 6738 |
Gene Name: | TROVE domain family member 2 |
Gene Aliases: |
RO60, RORNP, SSA2 |
Location: |
Chr.1:193059422-193091777 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TROVE2 | NM_001173524.1 | 1 | 252 | NP_001166995.1 |
| NR_033393.1 | 1 | 252 | ||
| XM_006711495.3 | 1 | 252 | XP_006711558.1 | |
| XM_006711496.3 | 1 | 79 | XP_006711559.1 | |
| XM_006711497.3 | 1 | 79 | XP_006711560.1 | |
| XM_011509922.2 | 1 | 79 | XP_011508224.1 | |
| XM_017002180.1 | 1 | 67 | XP_016857669.1 | |
| XM_017002181.1 | XP_016857670.1 | |||
| XM_017002182.1 | 1 | 67 | XP_016857671.1 | |
| XM_017002183.1 | 1 | 220 | XP_016857672.1 | |
| AK314594.1 | 1 | 39 | ||
| BI560207.1 | 1 | 44 | ||
| BX394323.2 | ||||
| CR936683.1 | 1 | 221 | ||
| CX761715.1 | 1 | 37 | ||
| DA518443.1 | 1 | 252 | ||
| S57803.1 |
Target Gene Details
Entrez Gene ID: | 51377 |
Gene Name: | ubiquitin C-terminal hydrolase L5 |
Gene Aliases: |
CGI-70, INO80R, UCH-L5, UCH37 |
Location: |
Chr.1:193012366-193060140 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCHL5 | NR_037607.1 | 1 | 435 | |
| XM_006711370.3 | XP_006711433.3 | |||
| XM_011509609.2 | 1 | 47 | XP_011507911.1 | |
| XM_017001434.1 | XP_016856923.1 | |||
| XM_017001435.1 | XP_016856924.1 | |||
| XM_017001438.1 | 1 | 47 | XP_016856927.1 | |
| XM_017001439.1 | 1 | 468 | XP_016856928.1 | |
| XM_017001440.1 | 1 | 432 | XP_016856929.1 | |
| XM_017001441.1 | 1 | 408 | XP_016856930.1 | |
| XM_017001444.1 | 1 | 417 | XP_016856933.1 | |
| AK301381.1 | 1 | 435 | ||
| BI548319.1 | 1 | 413 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv498n100 | Chr.1:192992568 - 193114915 on Build GRCh38 | Loss |
|
| nsv548658 | Chr.1:192394185 - 193552205 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map