Assay Details
Target Gene Details
Entrez Gene ID: | 1604 |
Gene Name: | CD55 molecule (Cromer blood group) |
Gene Aliases: |
CR, CROM, DAF, TC |
Location: |
Chr.1:207321472-207360966 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD55 | NM_000574.4 | NP_000565.1 | ||
| NM_001114752.2 | NP_001108224.1 | |||
| NM_001300902.1 | NP_001287831.1 | |||
| NM_001300903.1 | NP_001287832.1 | |||
| NM_001300904.1 | NP_001287833.1 | |||
| NR_125349.1 | ||||
| XM_017000467.1 | XP_016855956.1 | |||
| AB240566.1 | BAE97422.1 | |||
| AB240567.1 | BAE97423.1 | |||
| AB240568.1 | BAE97424.1 | |||
| AB240569.1 | BAE97425.1 | |||
| AB240570.1 | BAE97426.1 | |||
| AF052110.1 | ||||
| AK298549.1 | ||||
| AK300621.1 | ||||
| AK312448.1 | ||||
| AY055757.1 | AAL25832.1 | |||
| AY055758.1 | AAL25833.1 | |||
| AY055759.1 | 6 | 670 | AAL25834.1 | |
| AY055760.1 | AAL25835.1 | |||
| BC001288.1 | AAH01288.1 | |||
| BT007159.1 | AAP35823.1 | |||
| M15799.1 | AAA52167.1 | |||
| M30142.1 | AAA52168.1 | |||
| M31516.1 | AAA52169.1 | |||
| U88576.1 | AAB48622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002014 | Chr.1:207223904 - 207359600 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map