Assay Details
Target Gene Details
Entrez Gene ID: | 5519 |
Gene Name: | protein phosphatase 2 scaffold subunit Abeta |
Gene Aliases: |
PP2A-Abeta, PR65B |
Location: |
Chr.11:111694569-111766445 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R1B | XM_017017958.1 | XP_016873447.1 | ||
| XM_017017959.1 | XP_016873448.1 |
Target Gene Details
Entrez Gene ID: | 23235 |
Gene Name: | salt inducible kinase 2 |
Gene Aliases: |
LOH11CR1I, QIK, SIK-2, SNF1LK2 |
Location: |
Chr.11:111602391-111726917 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIK2 | NM_015191.2 | 10 | 1604 | NP_056006.1 |
| XM_017017417.1 | 10 | 1563 | XP_016872906.1 | |
| XM_017017418.1 | 8 | 1133 | XP_016872907.1 | |
| AB018324.3 | 10 | 1505 | ||
| AB096248.1 | 10 | 1510 | ||
| AK000396.1 | 10 | 1510 | ||
| AK291233.1 | 10 | 1539 | ||
| AL136764.2 | 10 | 1527 | CAB66698.1 | |
| BC078150.1 | 5 | 787 | AAH78150.1 | |
| BC113459.1 | 10 | 1461 | ||
| BC117183.1 | 10 | 1461 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832269 | Chr.11:111689546 - 111879947 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map