Assay Details
Target Gene Details
Entrez Gene ID: | 60509 |
Gene Name: | ATP/GTP binding protein like 5 |
Gene Aliases: |
CCP5, RP75 |
Location: |
Chr.2:27051423-27070622 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL5 | NM_001035507.2 | 2 | 299 | NP_001030584.1 |
| NM_021831.5 | 2 | 299 | NP_068603.4 | |
| NR_104246.1 | 2 | 299 | ||
| XM_005264477.2 | 2 | 299 | XP_005264534.1 | |
| XM_011533011.2 | 2 | 260 | XP_011531313.1 | |
| XM_011533012.2 | 3 | 465 | XP_011531314.1 | |
| XM_011533013.2 | 3 | 268 | XP_011531315.1 | |
| AK023398.1 | 2 | 254 | BAB14560.1 | |
| AK302091.1 | 2 | 230 | ||
| AL833844.1 | 2 | 241 | CAD38704.1 | |
| BC007415.2 | 2 | 241 | ||
| BC131498.1 | ||||
| CB155754.1 | 2 | 269 | ||
| DA901318.1 | 2 | 299 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6744n54 | Chr.2:26973431 - 27151141 on Build GRCh38 | Loss |
|
| nsv817673 | Chr.2:27037601 - 27163931 on Build GRCh38 | Gain |
|
| nsv455974 | Chr.2:26989775 - 27128411 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map