Assay Details
Target Gene Details
Entrez Gene ID: | 9650 |
Gene Name: | mitochondrial fission regulator 1 |
Gene Aliases: |
CHPPR, FAM54A2 |
Location: |
Chr.8:65643873-65771252 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTFR1 | XM_006716484.2 | XP_006716547.2 | ||
| XM_011517626.1 | XP_011515928.1 | |||
| XM_011517627.2 | XP_011515929.1 | |||
| XM_011517628.2 | XP_011515930.1 |
Target Gene Details
Entrez Gene ID: | 5150 |
Gene Name: | phosphodiesterase 7A |
Gene Aliases: |
HCP1, PDE7 |
Location: |
Chr.8:65714334-65841734 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE7A | NM_001242318.2 | 13 | 1607 | NP_001229247.1 |
| NM_002603.3 | 13 | 1456 | NP_002594.1 | |
| XM_011517540.2 | 14 | 1401 | XP_011515842.1 | |
| XM_017013538.1 | 13 | 1968 | XP_016869027.1 | |
| AK057586.1 | 1 | 1109 | ||
| AK290801.1 | 13 | 1415 | ||
| AK292680.1 | 13 | 1404 | ||
| BC126360.1 | 13 | 1448 | ||
| U67932.1 | 13 | 1459 | AAB65772.1 |
More Information
Set Membership: |
|

Genomic Map