Assay Details
Target Gene Details
Entrez Gene ID: | 84545 |
Gene Name: | mitochondrial ribosomal protein L43 |
Gene Aliases: |
L43mt, MRP-L43, bMRP36a |
Location: |
Chr.10:100977822-100987519 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL43 | NM_176792.2 | NP_789762.1 | ||
| NM_176793.1 | NP_789763.1 | |||
| NM_176794.1 | NP_789764.1 | |||
| XM_006718035.3 | XP_006718098.1 | |||
| AF086122.1 | ||||
| AK095556.1 | BAC04572.1 | |||
| AK300237.1 | ||||
| AW513604.1 | ||||
| BC041165.1 | AAH41165.1 | |||
| BC053373.1 | AAH53373.1 | |||
| BU072283.1 |
Target Gene Details
Entrez Gene ID: | 57715 |
Gene Name: | semaphorin 4G |
Gene Aliases: |
- |
Location: |
Chr.10:100969483-100985618 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA4G | NM_001203244.1 | NP_001190173.1 | ||
| NM_017893.3 | 14 | 2874 | NP_060363.2 | |
| XM_005270008.2 | 15 | 2948 | XP_005270065.1 | |
| AB046839.1 | 14 | 2568 | ||
| AK297431.1 | ||||
| BC051030.1 | 15 | 3261 | AAH51030.1 | |
| BC110382.1 | 14 | 2547 | ||
| BC128579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831965 | Chr.10:100916724 - 101082796 on Build GRCh38 | Loss |
|
| nsv517698 | Chr.10:100965896 - 101017113 on Build GRCh38 | Loss |
|
| nsv467443 | Chr.10:100929460 - 101007360 on Build GRCh38 | Loss |
|
| nsv552051 | Chr.10:100904892 - 100984619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map