Assay Details
Target Gene Details
Entrez Gene ID: | 55236 |
Gene Name: | ubiquitin like modifier activating enzyme 6 |
Gene Aliases: |
E1-L2, MOP-4, UBE1L2 |
Location: |
Chr.4:67615761-67701171 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA6 | NM_018227.5 | 33 | 4709 | NP_060697.4 |
| AK027020.1 | 3 | 1833 | ||
| AL832015.2 | 33 | 4678 | CAD89908.1 | |
| AL832458.2 | 33 | 4678 | CAD89959.1 | |
| AY359880.1 | 20 | 4030 | AAQ63403.1 |
More Information
Set Membership: |
|

Genomic Map