Assay Details
Target Gene Details
Entrez Gene ID: | 23524 |
Gene Name: | serine/arginine repetitive matrix 2 |
Gene Aliases: |
300-KD, CWF21, Cwc21, HSPC075, SRL300, SRm300 |
Location: |
Chr.16:2752329-2771412 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRRM2 | NM_016333.3 | NP_057417.3 | ||
| XM_005255226.1 | XP_005255283.1 | |||
| AB002322.3 | ||||
| AB016091.1 | BAA83717.1 | |||
| AB016092.1 | BAA83718.1 | |||
| AF201422.1 | AAF21439.1 | |||
| AI083775.1 | ||||
| AK025903.1 | ||||
| AL833672.1 | CAI46153.1 | |||
| BC009062.1 | AAH09062.1 | |||
| BC041155.1 | AAH41155.1 |
Target Gene Details
Entrez Gene ID: | 6923 |
Gene Name: | transcription elongation factor B subunit 2 |
Gene Aliases: |
ELOB, SIII |
Location: |
Chr.16:2771414-2777296 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEB2 | NM_007108.3 | NP_009039.1 | ||
| NM_207013.2 | NP_996896.1 | |||
| AI191983.1 | ||||
| BC013306.2 | AAH13306.1 | |||
| BM553320.1 | ||||
| BM700019.1 | ||||
| BM921799.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss |
|
| nsv471073 | Chr.16:2687070 - 2773569 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map