Assay Details
Target Gene Details
Entrez Gene ID: | 339221 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 7 |
Gene Aliases: |
ALK-SMase, E-NPP 7, NPP-7, NPP7 |
Location: |
Chr.17:79730919-79742926 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPP7 | NM_178543.4 | 6 | 1820 | NP_848638.3 |
| XM_011524737.1 | 5 | 1803 | XP_011523039.1 | |
| AK126250.1 | 6 | 2004 | BAC86504.1 | |
| AY230663.1 | 6 | 1803 | AAP69661.1 | |
| AY358622.1 | 6 | 1786 | AAQ88985.1 | |
| BC041453.1 | 6 | 1803 | AAH41453.2 |
More Information
Set Membership: |
|

Genomic Map